missing translation for 'onlineSavingsMsg'
Learn More

Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. p-7077879
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461260010 1 g
AC461260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461260010 Supplier Thermo Scientific Chemicals Supplier No. 461260010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78628-80-5
Molecular Formula C21H25N·HCl
Molecular Weight (g/mol) 327.89
InChI Key BWMISRWJRUSYEX-SZKNIZGXSA-N
Synonym terbinafine hydrochloride, terbinafine hcl, lamisil, terbinafine hydrochioride, lamisil krem, lamisil at, zabel, terbinafinum latin, terbinafina spanish, zimig
PubChem CID 5282481
ChEBI CHEBI:77614
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMILES CC(C)(C)C#CC=CCN(C)CC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 214.0°C
Loss on Drying 0.5% max. (1 g, 105)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information (5% in methanol) Clear solution
Formula Weight 327.89
Percent Purity 99%
Chemical Name or Material Terbinafine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only