missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terazosin Hydrochloride Dihydrate 98.0+%, TCI America™
$30.80 - $57.75
Chemical Identifiers
| CAS | 70024-40-7 |
|---|---|
| Molecular Formula | C19H30ClN5O6 |
| Molecular Weight (g/mol) | 459.928 |
| MDL Number | MFCD00941408 |
| InChI Key | NZMOFYDMGFQZLS-UHFFFAOYSA-N |
| Synonym | terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin |
| PubChem CID | 63016 |
| ChEBI | CHEBI:9446 |
| IUPAC Name | [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride |
| SMILES | COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2751100MG
|
TCI America
T2751100MG |
100 mg |
Each for $30.80
|
|
|||||
|
T27511G
|
TCI America
T27511G |
1 g |
Each for $57.75
|
|
|||||
Chemical Identifiers
| 70024-40-7 | |
| 459.928 | |
| NZMOFYDMGFQZLS-UHFFFAOYSA-N | |
| 63016 | |
| [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride |
| C19H30ClN5O6 | |
| MFCD00941408 | |
| terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin | |
| CHEBI:9446 | |
| COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl |
Specifications
| 70024-40-7 | |
| White | |
| MFCD00941408 | |
| terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin | |
| COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl | |
| 459.928 | |
| CHEBI:9446 | |
| ≥98.0% (HPLC,N) | |
| Terazosin Hydrochloride Dihydrate |
| 274°C | |
| C19H30ClN5O6 | |
| 100 mg | |
| NZMOFYDMGFQZLS-UHFFFAOYSA-N | |
| [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride | |
| 63016 | |
| 423.90 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TK8046000
TSCA : No