missing translation for 'onlineSavingsMsg'
Learn More

Terazosin Hydrochloride Dihydrate 98.0+%, TCI America™

Catalog No. T2751100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2751100MG 100 mg
T27511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2751100MG Supplier TCI America Supplier No. T2751100MG

Chemical Identifiers

CAS 70024-40-7
Molecular Formula C19H30ClN5O6
Molecular Weight (g/mol) 459.928
MDL Number MFCD00941408
InChI Key NZMOFYDMGFQZLS-UHFFFAOYSA-N
Synonym terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin
PubChem CID 63016
ChEBI CHEBI:9446
IUPAC Name [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl

Specifications

Melting Point 274°C
Color White
Quantity 100 mg
Formula Weight 423.90
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Terazosin Hydrochloride Dihydrate
RTECSNumber TK8046000
TSCA No