missing translation for 'onlineSavingsMsg'
Learn More

Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog No. AC460860050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460860050 5 g
AC460860010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460860050 Supplier Thermo Scientific Chemicals Supplier No. 460860050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70024-40-7 | ClH·2H2O | 459.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70024-40-7
Molecular Formula ClH·2H2O
Molecular Weight (g/mol) 459.92
InChI Key NZMOFYDMGFQZLS-UHFFFAOYSA-N
Synonym terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin
PubChem CID 63016
ChEBI CHEBI:9446
IUPAC Name [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl

Specifications

Melting Point 271.0°C to 274.0°C
Loss on Drying 9% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 459.92
Percent Purity 98%
Chemical Name or Material Terazosin hydrochloride dihydrate

RUO – Research Use Only