missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Terazosin Hydrochloride, ≥99%

Catalog No. AAJ62055MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62055MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62055MF Supplier Thermo Scientific™ Supplier No. J62055MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alpha-1 adrenoceptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage Ambient temperatures
For Use With (Application) Alpha-1 adrenoceptor antagonist
Assay ≥99%
CAS 63074-08-8
Molecular Formula C19H25N5O4·HCl
Formula Weight 423.90g/mol
Product Type Signal Transduction Reagent
Form Powder