Learn More
Wearwell™ Cleanroom Contamination Control Mat
Catalog No. 190341440
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
190341440 White
19151727 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 190341440 Supplier Wearwell™ Supplier No. 095.3X5WH
Sheets of tacky surface effectively pulls and traps dirt and dust off shoe soles
Cleanroom mats make it easy to guard against contamination and reduce facility maintenance.
  • Each mat has 30 numbered layers of tough film
  • Easy to use — pull off sheet when full and discard to expose new sheet
  • Adhesive backing holds mat securely in place
  • Tear resistant sheets are numbered to indicate when new mat is needed
  • Use on low-pile carpet, tile or concrete surfaces
Warranty
One year.
Specifications
Dimensions (L x W) 3 x 5 ft.
Color White