missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Tenax™ Porous Polymer Adsorbent, Matrix Tenax TA, 80-100 mesh

Catalog No. 111002480
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002480 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002480 Supplier MilliporeSigma™ Supelco™ Supplier No. 21009U

Tenax™ TA is suitable as an adsorbent used for air sampling, it was examined for its potential to form artifacts with Ozone at environmental concentrations, which was then quantitatively analysed using GC-MS analysis. It is also used in adsorbent columns suitable for recovering volatile compounds analysed by GC.

Porous polymer sorbents are inert, hydrophobic with large surface area. These sorbents retain volatile compounds, water and solvent vapours. This is advantageous if sample is from atmosphere with high content of water and solvent vapours. They are most suitable for trapping toxic agents with high molecular weight and non-volatile substances like pesticides. They are also suitable for sampling many compounds sorbed by charcoal and silica gel. Tenax™ TA is basically synthetically produced polymer based on the 2,6-diphenylene oxide is more efficient in the recovery of higher molecular weight compounds. It is suitable for both polar and nonpolar analytes.

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 10 g
Surface Area (Metric) ˜35 m2/g
Product Line Tenax
Density ˜0.29 g/mL (free fall density)

Tenax is a registered trademark of Buchem B.V.