missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ TenActiv™ Touchscreen Compatible Static and Cut-Resistant Gloves

Catalog No. 17400203
Change view
Click to view available options
Size:
0
1
2
5
6
7
8
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17400203 2
17400201 0
17400202 1
17400196 5
17400197 6
17400198 7
17400199 8
17400200 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17400203 Supplier Superior Glove™ Supplier No. S13TAPUCF12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cut-resistant gloves with polyurethane coating on palms and fingers, as well as touchscreen compatibility.

Perfect for electronics manufacturing, these gloves are made from a 13-gauge TenActiv™ blended yarn that provides ANSI Level A2 cut protection.

  • Carbon filament imparts anti-static and electrostatic dissipating (ESD) properties
  • Polyurethane palm coating for a strong, steady grip
  • Free of DMF (dimethylformamide)
  • Touchscreen compatible to move seamlessly between tasks

Specifications

Coating Material Polyurethane
Color Gray, Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material TenActiv Blended Yarn
Product Type Cut-resistant Gloves
Size 2
For Use With (Application) Electronics and Glass Manufacturing, Automotive Industry
Puncture Rating ANSI/ISEA 105-16 Level 3
Yarn Gauge 13
Length (English) Cuff 2 in.
Length (Metric) Cuff 5.1 cm
Show More Show Less