missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ TenActiv™ Knit Cut-Resistant Gloves, Polyurethane Coating

Catalog No. 17400204
Change view
Click to view available options
Size:
10
11
12
5
6
7
8
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17400204 5
17400205 6
17400206 7
17400207 8
17400208 9
17400209 10
17400210 11
17400211 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17400204 Supplier Superior Glove™ Supplier No. STAGHVPU5
Only null left

Knit, cut-resistant gloves with ANSI Cut Level A5 and polyurethane coating on palms and fingers.

A hi-vis knit shell gives these gloves an outstanding level of visibility on job sites, and their stronger-than-steel TenActiv™ yarn ensures ANSI Level A5 cut protection. Their shell has been made lint-free to avoid contaminants, and a polyurethane coating at the palms provides fingers with a tactile feel.

  • Made with high-strength TenActiv™ yarn that is stronger than steel on an equal weight basis
  • Polyurethane coating offers excellent dexterity and strong grip

Specifications

Coating Material Polyurethane
Color Hi-vis Yellow
Cut Resistance Rating ANSI A5
Material TenActiv Yarn
Size 5
For Use With (Application) Glass Manufacturing, Metal Fabrication, Automotive Industry
Puncture Rating ANSI/ISEA 105-16 Level 4
Yarn Gauge 13
Length (English) Cuff 2 in.
Length (Metric) Cuff 5.1 cm