Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Polyurethane Coating
Knit with high-tenacity composite yarns designed to meet a range of applications. Polyurethane coating offers a high coefficient of friction that is non-shedding.
Supplier: Superior Glove™ S13TAGPU7
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Withstands oils and solvents
Specifications
| Polyurethane | |
| ANSI A2 | |
| Cut-resistant Gloves | |
| Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |
| Gray | |
| Composite Fiber | |
| 7 | |
| Knit |