Learn More
Tenacity Mineral Collection
Explore tenacity—a mineral's ability to keep from breaking apart.
Supplier:  Geoscience Industries S45063
Catalog No. S45063
Only null left
Explore available promotions
Description
Description
Using terms like brittleness, elasticity, flexibility, malleability, and sectility, students will compare the tenacity of these six mineral specimens. Each measures approximately 1 1/2 x 1 1/2 in. and coded by number in a compartmented container.
Specifications
Specifications
Compare the tenacity (a mineral's ability to keep from breaking apart) of six mineral specimens
Six numbered specimens, ID list
Approx. 1 x 1.5 in.
Product Suggestions
Product Suggestions