Learn More
Thermo Scientific Chemicals Temsirolimus, 99+%
A Rapamycin derivative with anti-cancer activity that inhibits mTOR | CAS: 162635-04-3 | C56H87NO16
Supplier: Thermo Scientific Chemicals J63654MB
Description
Temsirolimus is used as a rapamycin derivative with anti-cancer activity that inhibits mTOR. Temsirolimus is an antiproliferative and antiangiogenic, the first-in-class mTOR inhibitor approved for the treatment of patients with advanced poor prognosis renal cell carcinoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 25 mg | |
| -30°C to -10°C | |
| ≥99% | |
| C56H87NO16 | |
| Signal Transduction Reagent |
| White to Off-white | |
| A Rapamycin derivative with anti-cancer activity that inhibits mTOR | |
| 162635-04-3 | |
| 1030.29g/mol | |
| Powder |
Chemical Identifiers
| 162635-04-3 | |
| C56H87NO16 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only