missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Temsirolimus, 99+%

Catalog No. AAJ63654MC
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63654MC 100 mg
AAJ63654MB 25 mg
AAJ63654MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63654MC Supplier Thermo Scientific Chemicals Supplier No. J63654MC

A Rapamycin derivative with anti-cancer activity that inhibits mTOR | CAS: 162635-04-3 | C56H87NO16

Temsirolimus is used as a rapamycin derivative with anti-cancer activity that inhibits mTOR. Temsirolimus is an antiproliferative and antiangiogenic, the first-in-class mTOR inhibitor approved for the treatment of patients with advanced poor prognosis renal cell carcinoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162635-04-3
Molecular Formula C56H87NO16

Specifications

Quantity 100 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A Rapamycin derivative with anti-cancer activity that inhibits mTOR
Formula Weight 1030.29g/mol
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
Recommended Storage Store at -20°C

RUO – Research Use Only