missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Wrist Length Cryogenic Gloves

Catalog No. 19276535
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19276535 Large
19276533 Small
19276534 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Catalog No. 19276535 Supplier Tempshield™ Supplier No. WRL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Wrist Length
Color Blue
Hand Specific
Product Type Cryogenic Gloves
Size Large
EN388 Standard Yes
For Use With (Application) Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
Series Cryo-Gloves™
Temperature (English) Min -320°F
Temperature (Metric) Min. -196°C
Length (English) 11.3 to 12.5 in.
Length (Metric) 29 to 32.5 cm
Show More Show Less