missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 6 SFR Hooded Coverall
Offer an extra layer of protection for primary fire-resistant clothing
Supplier: DuPont™ TM127SBU4X002500
Alert:
Should not be worn in presence of hazardous materials.
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Stormflaps.
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
| 4X-Large | |
| ProShield | |
| Single-use | |
| Elastic | |
| Standard | |
| Serged | |
| 32.5 in. | |
| Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities | |
| 100.33 cm | |
| 6.4 to 6.7 ft. |
| Unisex | |
| Blue | |
| Elastic | |
| Hooded | |
| Zipper | |
| Non-woven Wood Pulp, Polyester Fabric | |
| 128.9 to 137.79 cm | |
| 39.5 in. | |
| 32 in. | |
| 1.95 to 2.04 m |