missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 6 SFR Hooded Coverall

Catalog No. 19175272
Change view
Click to view available options
Color:
Blue
Light Blue
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19175272 Blue 6X-Large
19100386 Blue 5X-Large
296359904 Blue 4X-Large
191307801 Light Blue X-Large
191307800 Light Blue Large
191307803 Light Blue 3X-Large
29663156 Light Blue 2X-Large
29663153 Blue Medium
19175273 Blue 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Should not be worn in presence of hazardous materials.
Catalog No. 19175272 Supplier DuPont™ Supplier No. TM127SBU6X002500
Should not be worn in presence of hazardous materials.

Offer an extra layer of protection for primary fire-resistant clothing

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.

Compliance

Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.

Specifications

Size 6X-Large
Material ProShield
Color Blue
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper
Seam Type Serged
Description Non-woven Wood Pulp, Polyester Fabric
For Use With (Application) Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
Height (English) 6.9 to 7.1 ft.
Height (Metric) 2.1 to 2.16 m
Show More Show Less