missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 6 SFR Hooded Coverall
Offer an extra layer of protection for primary fire-resistant clothing
Supplier: DuPont™ TM127SBULG002500
Alert:
Should not be worn in presence of hazardous materials.
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Stormflaps.
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
| Large | |
| Light Blue | |
| Elastic | |
| Standard | |
| Serged | |
| 38.25 to 41.75 in. | |
| 97.15 to 106.04 cm | |
| Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities | |
| 88.9 cm | |
| 5.5 to 5.9 ft. |
| ProShield | |
| Elastic | |
| Hooded | |
| Zipper | |
| Non-woven Wood Pulp, Polyester Fabric | |
| 29 in. | |
| 73.66 cm | |
| 35 in. | |
| 25.75 in. | |
| 1.67 to 1.8 m |