Learn More
USA Scientific TempPlate RT optically clear film
Catalog No. NC9393791
Encompass
TempPlate RT optically clear polyester film, 2.0 mil thick, -40C to +120C, non-sterile, 100/box
Catalog No. NC9393791 Supplier USA Scientific Supplier No. 29782100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
TempPlate RT optically clear polyester film, 2.0 mil thick, -40C to +120C, non-sterile, 100/box