missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Temporary SecurAlert Adhesive Back Badge

Catalog No. 19103184
Change view
Click to view available options
Color:
Red on white
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19103184 Red on white
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103184 Supplier Brady™ Supplier No. 95690

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prevent temporary pass misuse. Brady™ Temporary SecurAlert Adhesive Back Badges are an ideal option for preventing unauthorized use of temporary visitor or contractor passes.

  • SecurAlert adhesive-back badges expire automatically to prevent automatic use
  • The expiration begins when the “front” badge is placed on the “back” badge
  • Expiration cannot be reversed once started

Specifications

Length (English) 2 in.
Color Red on white
Dimensions (L x W) 2 in. x 3 in.
Width (English) 3 in.