missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Temperature Chart Recorder Paper

Catalog No. 1399016A
Change view
Click to view available options
Temperature Range:
-200°C to 0°C
-40°C to +25°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range
1399016A -40°C to +25°C
13990214 -200°C to 0°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1399016A Supplier Thermo Scientific™ Supplier No. 6184

Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.

Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.

  • 6″ (15.2cm) diameter
  • For use in freestanding and panel-mounted chart recorders

Specifications

Type Chart Recorder Paper
Temperature Range -40°C to +25°C
For Use With (Equipment) Free-Standing and Panel-Mounted Chart Recorders
Monitoring Options Recorder
Static Holding Time 7 Days
Diameter (English) 6 in.
Diameter (Metric) 15.2 cm