missing translation for 'onlineSavingsMsg'
Learn More

Temozolomide 98.0+%, TCI America™

Catalog No. p-7137388
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2744500MG 500 mg
T27445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2744500MG Supplier TCI America Supplier No. T2744500MG

Chemical Identifiers

CAS 85622-93-1
Molecular Formula C6H6N6O2
Molecular Weight (g/mol) 194.154
MDL Number MFCD00866492
InChI Key BPEGJWRSRHCHSN-UHFFFAOYSA-N
Synonym temozolomide, methazolastone, temodal, temodar, temozolamide, temozolomidum, 3-methyl-4-oxo-3,4-dihydroimidazo 5,1-d 1,2,3,5 tetrazine-8-carboxamide, temozolomidum latin, temozolodida spanish, temozolodida
PubChem CID 5394
ChEBI CHEBI:72564
IUPAC Name 3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
SMILES CN1C(=O)N2C=NC(=C2N=N1)C(=O)N

Specifications

Melting Point 206°C
Quantity 500 mg
Formula Weight 194.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Temozolomide
RTECSNumber NJ5927050
TSCA No
Recommended Storage Refrigerator