missing translation for 'onlineSavingsMsg'
Learn More

Temozolomide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015687
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015687 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015687 Supplier Merck Emd Millipore Supplier No. PHR14371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.Temozolomide is an alkylating agent of the imidazotetrazine group. It is a chemotherapy drug, used in the treatment of anaplastic astrocytoma, glioblastoma multiforme and malignant metastatic melanoma, which works by arresting cancer cell growth.

Specifications

CAS 85622-93-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H6N6O2
MDL Number MFCD00866492
Synonym Temozolomide; 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide; 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5 ]tetrazinecarboxamide; 4-Methyl-5-oxo-2,3,4,6,8-pentazabicyclo[4.3.0 ]nona-2,7,9-triene-9-carboxamide; 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3 H)-one; NSC 362856
Type Certified Reference Material
Quantity 1 g
Formula Weight 194.15
Grade Certified Reference Material