Sciencell Research Laboratories Absolute Human Telomere Length Quantification qPCR Assay Kit 100 reactions
Catalog No. NC1503693
ScienCell's Absolute Human Telomere Length Quantification qPCR Assay Kit (AHTLQ) is designed to directly measure the average telomere length of a human cell population.
Catalog No.
NC1503693
Supplier
Sciencell Research LaboratoriesSupplier No.
8918
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ScienCell's Absolute Human Telomere Length Quantification qPCR Assay Kit (AHTLQ) is designed to directly measure the average telomere length of a human cell population.