missing translation for 'onlineSavingsMsg'
Learn More

Telmisartan 98.0+%, TCI America™

Catalog No. p-7137456
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T28611G 1 g
T28615G 5 g
2 options

Catalog No. T28611G

Supplier: TCI America T28611G

Only null left

Chemical Identifiers

CAS 144701-48-4
Molecular Formula C33H30N4O2
Molecular Weight (g/mol) 514.629
MDL Number MFCD00918125
InChI Key RMMXLENWKUUMAY-UHFFFAOYSA-N
Synonym telmisartan, micardis, pritor, bibr 277, kinzalmono, bibr 277se, 4'-1,7'-dimethyl-2'-propyl-1h,3'h-2,5'-bibenzo d imidazol-3'-yl methyl-1,1'-biphenyl-2-carboxylic acid, bibr-277, targit, telmisartan inn
PubChem CID 65999
ChEBI CHEBI:9434
IUPAC Name 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoic acid
SMILES CCCC1=NC2=C(C=C(C=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C(=O)O)C5=NC6=CC=CC=C6N5C)C

Specifications

Melting Point 269°C
Color White
Quantity 1 g
Formula Weight 514.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Telmisartan
RTECSNumber DV2037500
TSCA No