missing translation for 'onlineSavingsMsg'
Learn More

Tellurium shot, 2-5mm (0.08-0.20in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4408180
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4221306 5 g
AA4221314 25 g
AA4221322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4221306 Supplier Thermo Scientific Chemicals Supplier No. 04221306
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Color Silver-White
Physical Form Shot
Quantity 5 g
Assay Percent Range 99.9999% (Metals basis)
Odor Odorless
Appearance Silvery-white shot
Chemical Name or Material Tellurium
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
EINECSNumber 236-813-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only