missing translation for 'onlineSavingsMsg'
Learn More

Tellurium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4345018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4345018 50 g
AA4345009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4345018 Supplier Thermo Scientific Chemicals Supplier No. 04345018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Quantity 50 g
Mesh Size −325 Mesh
Odor Odorless
Form Powder
UN Number UN3288
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Tellurium powder
Hazard Category H301
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 236-813-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only