missing translation for 'onlineSavingsMsg'
Learn More

Tellurium powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4824977
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1107509 10 g
AA1107518 50 g
AA1107530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1107509 Supplier Thermo Scientific Chemicals Supplier No. 01107509
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Color Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Odor Odorless
Appearance Gray powder
Chemical Name or Material Tellurium
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 236-813-4
RTECSNumber WY2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only