missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) Oxide 98.0+%, TCI America™

Catalog No. T325425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T325425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T325425G Supplier TCI America Supplier No. T325425G

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Synonym tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
PubChem CID 62638
IUPAC Name (oxo-λ⁴-tellanylidene)oxidane
SMILES O=[Te]=O

Specifications

Melting Point 733°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 159.60
Percent Purity ≥98.0% (W)
Chemical Name or Material Tellurium(IV) Oxide
EINECSNumber (1)-0729
RTECSNumber WY2675000
TSCA Yes