missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) oxide, 99+%

Catalog No. AC222590500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222590500 50 g Glass bottle
AC222592500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222590500 Supplier Thermo Scientific Chemicals Supplier No. 222590500

CAS: 7446-07-3 | O2Se | 110.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name (oxo-λ⁴-selanylidene)oxidane
SMILES O=[Se]=O

Specifications

Boiling Point 1245°C
Melting Point 733°C
Color White to Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99+%
Linear Formula TeO2
Merck Index 15, 9265
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. acids and alkalies,slightly soluble in diluted acids
Formula Weight 159.6
Percent Purity ≥99%
Packaging Glass bottle
Density 5.67g/mL
Chemical Name or Material Tellurium(IV) oxide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if inhaled.
May damage fertility or the unborn child.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
EINECSNumber 231-193-1
RTECSNumber WY2675000
TSCA TSCA

RUO – Research Use Only