missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) oxide, 99.995% (metals basis)

CAS: 7446-07-3 | O2Te | 159.60 g/mol

Supplier:  Thermo Scientific Chemicals 01182118

 View more versions of this product

Catalog No. AA1182118


Only null left
Explore available promotions

Description

Description

Tellurium(IV) oxide is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tellurium(IV) oxide
7446-07-3
Powder
99.995% metal basis
MFCD00011263
Soluble in strong acids and alkali. Insoluble in water.
O=[Te]=O
159.60
159.6
5.8
733°C
White to Yellow
50 g
O2Te
tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium dioxide
62638
Odorless

Chemical Identifiers

7446-07-3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c

H317-H332-H360

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-193-1

RTECSNumber : WY2675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only