missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) oxide, 99.99% (metals basis)

Catalog No. p-4408182
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8781314 25 g
AA8781322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8781314 Supplier Thermo Scientific Chemicals Supplier No. 08781314
Only null left

CAS: 7446-07-3 | O2Te | 159.60 g/mol

Tellurium(IV) oxide is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Synonym tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
PubChem CID 62638
IUPAC Name (oxo-λ⁴-tellanylidene)oxidane
SMILES O=[Te]=O

Specifications

Melting Point 733°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in strong acids and alkali. Insoluble in water.
Formula Weight 159.6
Density 5.8 g/mL
Chemical Name or Material Tellurium(IV) oxide
Hazard Category H317-H332-H360
Precautionary Statement P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-193-1
RTECSNumber WY2675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only