missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) chloride, 99%

CAS: 10026-07-0 | Cl4Te | 269.4 g/mol

$183.05 - $183.05

Chemical Identifiers

CAS 10026-07-0
Molecular Formula Cl4Te
Molecular Weight (g/mol) 269.4
MDL Number MFCD00011262
InChI Key SWLJJEFSPJCUBD-UHFFFAOYSA-N
Synonym tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
PubChem CID 61443
SMILES Cl[Te](Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC194740250
SDS View Documents
Thermo Scientific Chemicals
194740250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10026-07-0
269.4
SWLJJEFSPJCUBD-UHFFFAOYSA-N
61443
Cl4Te
MFCD00011262
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
Cl[Te](Cl)(Cl)Cl

Specifications

380.0°C
10026-07-0
Powder
98.5% min. (ex Cl) (Argentometry)
TeCl4
09,443; 10,377; 12,457; 16,316
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
SWLJJEFSPJCUBD-UHFFFAOYSA-N
269.4
269.4
Glass bottle
3.2600g/mL
224.0°C
White to Yellow
25 g
Cl4Te
MFCD00011262
15, 9268
Solubility in water: may decompose. Other solubilities: forming teo2 and hcl, soluble in absolute alcohol and toluene
Cl[Te](Cl)(Cl)Cl
61443
99%
3.26
Tellurium(IV) chloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-055-6

RTECSNumber : WY2635000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only