missing translation for 'onlineSavingsMsg'
Learn More

Tellurium broken ingot, Puratronic™, 99.9999+% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049052
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1260714 25 g
AA1260722 100 g
AA1260736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1260714 Supplier Thermo Scientific Chemicals Supplier No. 01260714
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Quantity 25 g
Odor Odorless
Form Ingot
Percent Purity ≥99.9999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tellurium broken ingot
Hazard Category H301
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
EINECSNumber 236-813-4
RTECSNumber WY2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only