missing translation for 'onlineSavingsMsg'
Learn More

Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific™

Numéro de catalogue. p-4228283
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC437010250 25g Glass bottle
AC437011000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC437010250 Fournisseur Thermo Scientific Chemicals Code fournisseur 437010250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis

Identifiants chimiques

CAS 13494-80-9
Formule moléculaire Te
Poids moléculaire (g/mol) 127.6
Clé InChI PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonyme tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
Nom de l’IUPAC tellurium
SOURIRES [Te]

Spécifications

Nom ou substance chimique Tellurium
Quantité 25g
Fourchette de pourcentages d’essai 99.999%
Indice de Merck 15,9263
Renseignements sur la solubilité Solubility in water: insoluble. Other solubilities: soluble in h2so4,hno3 and koh,insoluble in benzene and carbon disulfide
Poids de formule 127.6
Pourcentage de pureté 99.999%
Niveau Trace Metal Basis
Emballage Glass bottle
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
EINECSNumber 236-813-4

RUO – Research Use Only