missing translation for 'onlineSavingsMsg'
Learn More

Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 437011000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC437011000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
Specifications

Chemical Identifiers

13494-80-9
127.6
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
CHEBI:30452
[Te]
Te
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium

Specifications

Tellurium
100g
Te
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
127.6
Trace Metal Basis
13494-80-9
99.999%
15,9263
Solubility in water: insoluble. Other solubilities: soluble in h2so4,hno3 and koh,insoluble in benzene and carbon disulfide
[Te]
127.6
CHEBI:30452
99.999%
Glass bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

GHS Signal Word: Warning

EINECSNumber : 236-813-4

SDS
Documents

Documents