missing translation for 'onlineSavingsMsg'
Learn More

Telluric Acid 99.0+%, TCI America™

Catalog No. T325325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T325325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T325325G Supplier TCI America Supplier No. T325325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7803-68-1
Molecular Formula H6O6Te
Molecular Weight (g/mol) 229.642
MDL Number MFCD00011609
InChI Key FXADMRZICBQPQY-UHFFFAOYSA-N
Synonym orthotelluric acid, telluric vi acid, tellurium hydroxide, telluric acid h6teo6, unii-we1kzr49wu, ccris 9168, we1kzr49wu, telluricacid, tellursaure, orthotellursaeure
PubChem CID 62686
ChEBI CHEBI:30461
SMILES O[Te](O)(O)(O)(O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3284
Formula Weight 229.64
Percent Purity ≥99.0% (T)
Chemical Name or Material Telluric Acid
EINECSNumber (1)-0729
RTECSNumber WY2350000
TSCA Yes