missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Telithromycin, 95%

CAS: 191114-48-4 | C43H65N5O10 | 812.018 g/mol

Supplier:  Thermo Scientific Chemicals J66820MC

Catalog No. AAJ66820MC

Only null left
Explore available promotions

Description

Description

Telithromycin is used to treat respiratory infections. Telithromycin prevents bacteria from growing, by interfering with their protein synthesis. Telithromycin binds to the subunit 50S of the bacterial ribosome, and blocks the progression of the growing polypeptide chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Telithromycin
176°C to 188°C
>110°C (230°F)
C43H65N5O10
100 mg
telithromycin, 1r,2r,4r,6s,7r,8r,10r,13r,14s-7-2s,3r,4s,6r-4-dimethylamino-3-hydroxy-6-methyloxan-2-yl oxy-13-ethyl-6-methoxy-2,4,6,8,10,14-hexamethyl-17-4-4-pyridin-3-ylimidazol-1-yl butyl-12,15-dioxa-17-azabicyclo 12.3.0 heptadecane-3,9,11,16-tetrone
LJVAJPDWBABPEJ-PDDWODRRSA-N
(1R,2R,4R,6R,7R,8R,10R,13R)-7-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-13-ethyl-6-methoxy-2,4,6,8,10,14-hexamethyl-17-[4-(4-pyridin-3-ylimidazol-1-yl)butyl]-12,15-dioxa-17-azabicyclo[12.3.0]heptadecane-3,9,11,16-tetrone
122130735
95%
191114-48-4
966°C
Plastic bottle
MFCD04117983
UN3077
Sparingly soluble in water
CCC1C2(C(C(C(=O)C(CC(C(C(C(=O)C(C(=O)O1)C)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)N(C(=O)O2)CCCCN4C=C(N=C4)C5=CN=CC=C5)C
812.018
812
Powder

Chemical Identifiers

191114-48-4
812.018
LJVAJPDWBABPEJ-PDDWODRRSA-N
122130735
CCC1C2(C(C(C(=O)C(CC(C(C(C(=O)C(C(=O)O1)C)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)N(C(=O)O2)CCCCN4C=C(N=C4)C5=CN=CC=C5)C
C43H65N5O10
MFCD04117983
telithromycin, 1r,2r,4r,6s,7r,8r,10r,13r,14s-7-2s,3r,4s,6r-4-dimethylamino-3-hydroxy-6-methyloxan-2-yl oxy-13-ethyl-6-methoxy-2,4,6,8,10,14-hexamethyl-17-4-4-pyridin-3-ylimidazol-1-yl butyl-12,15-dioxa-17-azabicyclo 12.3.0 heptadecane-3,9,11,16-tetrone
(1R,2R,4R,6R,7R,8R,10R,13R)-7-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-13-ethyl-6-methoxy-2,4,6,8,10,14-hexamethyl-17-[4-(4-pyridin-3-ylimidazol-1-yl)butyl]-12,15-dioxa-17-azabicyclo[12.3.0]heptadecane-3,9,11,16-tetrone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

RTECSNumber : KF4674500

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only