missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Telegraph Key

Catalog No. S15768
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S15768 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15768 Supplier United Scientific™ Supplier No. TGKY01

Can be used in a variety of electricity demonstrations including Morse Code.

  • Telegraph key, mounted on a solid plastic base.
  • Measures 11 x 5 cm, with two screw-type connecting terminals.

Specifications

Grade 5 to 12
Includes Telegraph Key
Class Size 1 Lab Group
Product Type Electricity Education

Warning: Cancer and reproductive harm