missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ Tegrant ThermoSafe LabMailer Absorbent Sheets

Catalog No. 03600161
Change view
Click to view available options
Capacity:
22.5 mL of blood/sheet
360 mL of water/90 mL blood/sheet
Diameter (Metric) Interior:
15.2 x 15.2 cm
30.4 x 30.4 cm
7.6 x 7.6 cm
Diameter (English) Interior:
3 x 3 in.
6 x 6 in.
12 x 12 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Diameter (Metric) Interior Diameter (English) Interior
03600161 360 mL of water/90 mL blood/sheet 30.4 x 30.4 cm 12 x 12 in.
0353162 22.5 mL of blood/sheet 7.6 x 7.6 cm 3 x 3 in.
03600160 360 mL of water/90 mL blood/sheet 15.2 x 15.2 cm 6 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03600161 Supplier Sonoco™ Supplier No. ZORB1212

Lightweight, economical lab mailer shippers

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Polystyrene foam construction saves postage
  • Reusable and recyclable

Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).

Specifications

Length (English) Exterior 12 in.
For Use With (Equipment) Provide maximum breakage protection for known non-hazardous materials.
Applications For protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
Height (English) Exterior 12 in.
Diameter (English) Interior 12 x 12 in.
Diameter (Metric) Interior 30.4 x 30.4 cm
Capacity 360 mL of water/90 mL blood/sheet
Material Lightweight Polystyrene Foam