Learn More
Eisco™ Tectonics Model
Catalog No. S012319
Change view
Click to view available options
Dimensions (L x W):
21 x 18.5 in.
Catalog No. Dimensions (L x W)
S012319 21 x 18.5 in.
1 options
Catalog No. S012319 Supplier Eisco™ Supplier No. BD0090
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tectonics Model showing detailed representations of the Earth's layers, including both active and passive margins, oceanic ridges with rift valley, Benioff plane, oceanic trenches, continental shelf and slopes.
  • Visual representation to communication the importance of plate movements in relation to volcanism and seismicity
  • Each specific feature is labeled for easy identification
  • Crafted by subject matter experts
  • Features vibrant, hand-painted details using eco-friendly paint
  • Designed as a visual learning aid for introducing and exploring various geological concepts
  • Oceanic floor spreading is also well characterized to illustrate the development of a rift into a new structure
Specifications
Product Type Tectonics Model Set
Dimensions (L x W) 21 x 18.5 in.