missing translation for 'onlineSavingsMsg'
Learn More

Tebbe reagent, 0.5M solution in toluene, AcroSeal™

CAS: 67719-69-1 | C13H18AlClTi

Supplier:  Thermo Scientific Chemicals 426061000

Catalog No. AC426061000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination), Lewis acid
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only