Learn More
Therapak™ TearTech™ Tube Shuttles
Separates tubes to prevent contact
$1174.09 - $1174.09
Specifications
Applications Blood and urine tube mailings
View More Specs
Products 1
Catalog Number Mfr. No. Closure Type Quantity Price Quantity  
Catalog Number Mfr. No. Closure Type Quantity Price Quantity  
22-130-038
View Documents
Therapak™
15540
Zipper Up to 8 tubes
Case of 500 Each for $1,174.09
Only null left
Description
Description
  • Leak-tight construction for increased safety during transport
  • Numbered tube slots for easy reference
  • Two sizes available to hold six or eight blood or urine tubes (12–16mm dia.)
  • Six-tube shuttle features a TearTech closure that folds down for a permanent seal, yet provides a natural perforation for easy opening at the lab
  • Eight tube shuttle features a zip-style closure
  • Shuttles can be rolled or folded and placed inside a secondary watertight container
Specifications
Specifications
Blood and urine tube mailings
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'