Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cassettes DFS Item
For use with TE62 Standard Transfer Tank with Cooling Chamber
$307.00 - $307.00
Specifications
Includes 1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity  
03-500-283 DFS Item
View Documents
Hoefer™
HFRTE24
TE 22 Mini Tank Transfer Unit Gel Cassette
Pack for $307.00
Only null left
Description
Description
  • Accessories and Replacement Parts for TE62
Specifications
Specifications
1 cassette assembly, 2 Foam sponges 3mm thick and 1 Foam sponge 6mm thick
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'