missing translation for 'onlineSavingsMsg'
Learn More

TBTU 98.0+%, TCI America™

Catalog No. b16585g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16585G 5 g
B165825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16585G Supplier TCI America Supplier No. B16585G
Only null left

Specifications

Purity ≥98.0% (HPLC)
CAS 125700-67-6
Format Crystal-Powder at 20°C
Product Type TBTU [Coupling Reagent for Peptide]
Quantity 5 g