missing translation for 'onlineSavingsMsg'
Learn More

General Data Healthcare™ SHUR/Stain™ Manual Stainers

Manual stain rack with individual lids for use during extended inactivity

Supplier:  General Data Healthcare™ SS3L

 View more versions of this product

Catalog No. 22048001


Only null left

Description

Description

  • Metal extruded frames inter-lock for back-to-back connection of multiple racks
  • See-through lexan cover included
  • Plastic dishes are solvent resistant and include lids for limiting evaporation
  • With digital timer and label strips to record reagent expiration dates

SS-3L:

  • 3 positions
  • Includes two white 250mL wells (SS-WLW), one green solvent-resistant 250mL well (SS-WLG), one 24-position plastic slide carrier (SS-SRG-24)
  • See-through Lexan™ cover
  • Three 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 8 x 7 in. (160 x 203 x 178mm)

SS-6L:

  • 6 positions
  • Includes four white 250mL wells (SS-WLW), two green solvent-resistant 250mL well (SS-WLG), one 24-poisition plastic slide carrier (SS-SRG-24)
  • See-through plastic lid
  • Six 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 15 x 6 in. (160 x 375 x 156mm)

SS-12L:

  • 12 positions
  • Includes eight white 250mL wells (SS-WLW), four green solvent-resistant 250mL well (SS-WLG), one 24-poisition plastic slide carrier (SS-SRG-24)
  • See-through plastic lid
  • Twelve 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 28 x 6 in. (160 x 718 x 156mm)
Specifications

Chemical Identifiers

Plastic
6 in.
16 cm

Specifications

Plastic
7 x 8 in. (17.8 x 20.3cm)
7 in.
6 in.
20.3 cm
3
17.8 cm
8 in.
16 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions