missing translation for 'onlineSavingsMsg'
Learn More

General Data Healthcare™ Flotation Work Station Bath

Catalog No. 22047236
Change view
Click to view available options
Amperage:
1.75 A
2.5 A
3.5 A
5 A
Type:
Flotation Work Station
Tissue Flotation Bath
Model:
FWS-L
FWS-S
TFB-L
TFB-S
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Amperage Type Model
22047236 1.75 A Tissue Flotation Bath TFB-S
22047230 5 A Flotation Work Station FWS-L
22047234 3.5 A Tissue Flotation Bath TFB-L
22047232 2.5 A Flotation Work Station FWS-S
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22047236 Supplier General Data Healthcare™ Supplier No. TFBS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General Data's baths provide optimum specimen viewing and the dependability and safety of up-to-date temperature controlled electronics...making tissue preparation easier, safer and more efficient.

Ergonomically designed to provide reliability with advanced digital microprocessor control over temperature protection for preparation of paraffin embedded tissue sections

Combined with General Data’s histology consumable and reagents, these baths ensure whole ribbon sections are properly collected without cracking or over-expansion

Basic Bath

  • Bath control panel features a keypad and two LED displays
  • Arrow keys let you set and display water temperature from ambient up to 65°C (149°F)
  • Microprocessor-controlled components heat bath quickly and evenly (ambient to 65°C [149°F])
  • Second LED display switches between actual water temperature and time of day
  • Magnetic, noncontact switch functions flawlessly for years
Workstation
  • Includes everything in the basic bath, and also integrates a HISTO/Orientator™ heating block and slide dryer
  • The 2 × 2 in. (5 × 5cm) heating block can be used after removing slide from the bath to eliminate any remaining bubbles in the tissue
  • The 2 × 8 in. (5 × 20cm) slide dryer can dry several slides simultaneously
  • Both heating block and slide dryer located above control panel so as not to interfere with bath

 

Order Info

Accessories are available (See Cat. Nos. 22-047-239, -241, -242 and 22-044-630).

Specifications

Controller Type Microprocessor
Certifications/Compliance UL, CSA, CE
Type Tissue Flotation Bath
Display LED
Includes 8 x 8 x 2.25 in. D Dish removable Glass bowl with built-in clock and LED light array
Model TFB-S
Dimensions (L x W x H) 18.5 x 14 x 5.4 in. (47 x 36 x 14 cm)
Length (English) 18.5 in.
Length (Metric) 47 cm
Width (English) 14 in.
Width (Metric) 36 cm
Height (English) 5.4 in.
Height (Metric) 14 cm
Weight (English) 11 lb.
Weight (Metric) 6 kg
Electrical Requirements 100/120 V 50/60 Hz
Voltage 100/120 V
Amperage 1.75 A
Frequency 50/60 Hz
Show More Show Less