missing translation for 'onlineSavingsMsg'
Learn More

General Data Healthcare™ Flotation Work Station Bath

General Data's baths provide optimum specimen viewing and the dependability and safety of up-to-date temperature controlled electronics...making tissue preparation easier, safer and more efficient.

Supplier:  General Data Healthcare™ FWSS

 View more versions of this product

Catalog No. 22047232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Ergonomically designed to provide reliability with advanced digital microprocessor control over temperature protection for preparation of paraffin embedded tissue sections

Combined with General Data’s histology consumable and reagents, these baths ensure whole ribbon sections are properly collected without cracking or over-expansion

Basic Bath

  • Bath control panel features a keypad and two LED displays
  • Arrow keys let you set and display water temperature from ambient up to 65°C (149°F)
  • Microprocessor-controlled components heat bath quickly and evenly (ambient to 65°C [149°F])
  • Second LED display switches between actual water temperature and time of day
  • Magnetic, noncontact switch functions flawlessly for years
Workstation
  • Includes everything in the basic bath, and also integrates a HISTO/Orientator™ heating block and slide dryer
  • The 2 × 2 in. (5 × 5cm) heating block can be used after removing slide from the bath to eliminate any remaining bubbles in the tissue
  • The 2 × 8 in. (5 × 20cm) slide dryer can dry several slides simultaneously
  • Both heating block and slide dryer located above control panel so as not to interfere with bath

 

Order Info

Accessories are available (See Cat. Nos. 22-047-239, -241, -242 and 22-044-630).

Specifications

Chemical Identifiers

Microprocessor
LED
18.5 in.
14 in.
5.4 in.
11 lb.
100/120 V 50/60 Hz
50/60 Hz
UL, CSA, CE
18.5 x 14 x 5.4 in. (47 x 36 x 14 cm)
47 cm
36 cm
14 cm
6 kg
100/120 V

Specifications

Microprocessor
Flotation Work Station
8 x 8 x 2.25 in. D Dish removable Glass bowl with built-in LED light array, 2 x 6 in. slide dryer, 2 x 2 in. HISTO/orientator
18.5 x 14 x 5.4 in. (47 x 36 x 14 cm)
47 cm
36 cm
14 cm
6 kg
100/120 V
50/60 Hz
UL, CSA, CE
LED
FWS-S
18.5 in.
14 in.
5.4 in.
11 lb.
100/120 V 50/60 Hz
2.5 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions