Learn More
Tasco™ TB-2 Capmount Visor Carrier
Catalog No. 296361307
Change view
Click to view available options
TB-2 Capmount Visor Carrier
Catalog No.
296361307 TB-2 Capmount Visor Carrier
1 options
Catalog No. 296361307 Supplier Tasco™ Supplier No. DTC125000A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
TB-2 Capmount Visor Carrier with A-10 universal blades
  • Fits into all Tasco™ cap-mounted earmuffs
  • Adjustable to fit all cap-style hard hat brims
  • Easily raised and lowered
  • Holds .040 and .060 windows from 12 in. to 20 ft. in length and metal mesh visor
Specifications
For Use With (Equipment) Hard Hats
Product Type Hard Hat Visor Carrier
Size Universal
Ventilated Not Ventilated