missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tauroursodeoxycholic Acid, Sodium Salt, Calbiochem™,

Catalog No. 5805491GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5805491GM 1 g Glass Bottle
5805495GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5805491GM Supplier MilliporeSigma™ Supplier No. 5805491GM

Chemical Identifiers

CAS 14605-22-2
Molecular Formula C26H45NO6S
Molecular Weight (g/mol) 499.71
MDL Number MFCD00065451
InChI Key BHTRKEVKTKCXOH-AYSJQVDDSA-N
Synonym Sodium Tauroursodeoxycholate
PubChem CID 9848818
ChEBI CHEBI:80774
IUPAC Name 2-[(4R)-4-[(1R,4S,5aS,7R,9aS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonic acid
SMILES C[C@H](CCC(=O)NCCS(O)(=O)=O)[C@H]1CCC2C3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)C3CC[C@]12C

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥97% (TLC)