missing translation for 'onlineSavingsMsg'
Learn More

Taurine, 99%

Catalog No. AC166540250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166540250 25 g Plastic bottle
AC166541000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166540250 Supplier Thermo Scientific Chemicals Supplier No. 166540250

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
PubChem CID 1123
ChEBI CHEBI:15891
IUPAC Name 2-aminoethanesulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Melting Point 300°C
Color White
Loss on Drying 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CH2SO3H
Quantity 25 g
Beilstein 04, 528
Merck Index 15, 9211
Solubility Information Solubility in water: 65g/L (12°C). Other solubilities: insoluble in absolute ethanol,diethyl ether,soluble in dmso
Formula Weight 125.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Taurine, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-483-8
RTECSNumber WX0175000
TSCA TSCA

RUO – Research Use Only